Glass Beads, SiLibeads®
Catalog # LINR7503-20KG
Supplier: SIGMUND LINDNER
Specifications
- Ø:0,75 - 1,00 mm
- Description:Glass beads
- Pk:20 kg
Specifications
About this item
Aluminium-borosilicate glass.
SiLibeads® Type SL glass beads used as grinding media for horizontal and vertical bead mills.