Order Entry
Finland
ContactUsLinkComponent
Anti-TRPC7 Mouse Monoclonal Antibody (Unconjugated) [clone: S64A-36]
Anti-TRPC7 Mouse Monoclonal Antibody (Unconjugated) [clone: S64A-36]
Catalog # ROCK200-301-G56
undefined
Anti-TRPC7 Mouse Monoclonal Antibody (Unconjugated) [clone: S64A-36]
Catalog # ROCK200-301-G56

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    transient receptor potential cation channel, subfamily C, member 7
  • Antigen symbol:
    TRPC7
  • Clonality:
    Monoclonal
  • Clone:
    S64A-36
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    57113
  • Antigen synonyms:
    TRP7
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    TrpC7 Antibody was produced in mice by repeated immunizations raised against a synthetic peptide corresponding to a near c-terminus region of human TrpC7.
  • Purification:
    Protein G purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Anti-TrpC7 Antibody is suitable for use in WB, IHC and IP. Expect a band approximately ~100kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: TRPC7
Clonality: Monoclonal
Clone: S64A-36
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat, Human