Anti-DLG4 Mouse Monoclonal Antibody (Unconjugated) [clone: 6G6]
Catalog # ROCK200-301-G32
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Discs, large homolog 4 (Drosophila)
- Antigen symbol:DLG4
- Clonality:Monoclonal
- Clone:6G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Bovine,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:1742
- Antigen synonyms:SAP-90|SAP90|PSD95
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:PSD95 Antibody was produced in mice by repeated immunizations raised against recombinant rat PSD-95.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-PSD-95 Antibody is suitable for use in WB, IHC, Immuno-gold EM, and IP. Expect a band approximately ~95-110kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: DLG4
Clonality: Monoclonal
Clone: 6G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Bovine, Rat