Anti-IgG1 Goat Polyclonal Antibody (ATTO 550)
Catalog # ROCK610-154-040
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG1
- Clonality:Polyclonal
- Conjugation:ATTO 550
- Host:Goat
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Western blot:Yes
- ChIP:Yes
- Tested applications:Fluoresence ELISA, Fluorescence Resonance Energy Transfer, Stimulated Emission Depletion Microscopy
- Size:500 µg
- Pk:500 µG
Specifications
About this item
The emission spectra for this ATTO conjugate matches the principle output wavelengths of most common fluorescence instrumentation.
Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 500 µl
Type: Secondary
Antigen: IgG1 γ
Clonality: Polyclonal
Clone:
Conjugation: ATTO 550
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse