Anti-HAUS8 Rabbit Polyclonal Antibody
ROCK600-401-C53
- Antibody type:Primary
- Antigen name:HAUS augmin-like complex subunit 8
- Antigen symbol:HAUS8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:93323
- Antigen synonyms:DGT4|HICE1|NY-SAR-48
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-Hice1 Antibody was prepared by repeated immunizations with a synthetic peptide corresponding to the region of amino acids containing serine 70 of human Hice1.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Hice1 antibody has been tested for use in ELISA and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 44.9 kDa in size corresponding to human Hice1 Protein by western blotting in the appropriate cell lysate.
Recommended Dilutions: ELISA: 1:30,000-1:45000; Western Blot: 0.5 µg/ml;
Type: Primary
Antigen: HAUS8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human