Anti-ATG13 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-C50
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:autophagy related 13
- Antigen symbol:ATG13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:9776
- Antigen synonyms:KIAA0652|PARATARG8
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This affinity purified antibody was prepared by repeated immunizations with a synthetic peptide corresponding to the S318 region of ATG13.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This affinity purified antibody has been tested for use in ELISA and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 56.6 kDa in size corresponding to human ATG13 Protein by western blotting.
Recommended Dilutions: ELISA: 1:220000-1:250000; Western Blot: 1:1000;
Type: Primary
Antigen: ATG13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human