Order Entry
Finland
ContactUsLinkComponent
 
Anti-SMAD3 Rabbit Polyclonal Antibody
  ROCK600-401-C48
undefined
Anti-SMAD3 Rabbit Polyclonal Antibody
  ROCK600-401-C48

 

  • Antibody type:
    Primary
  • Antigen symbol:
    SMAD family member 3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Internal
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    4088
  • Antigen synonyms:
    JV15-2|LDS3|HsT17436|HSPC193|LDS1C|MADH3
  • Modification:
    phospho-Thr179
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    Anti-SMAD3 pT179 antibody was prepared by repeated immunizations with a synthetic peptide corresponding to an internal region of human Smad3 Protein surrounding amino acid residue 179.
  • Purification:
    affinity-purified
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Anti-SMAD3 pT179 has been tested for use in ELISA.

Recommended Dilutions: ELISA: 1:15000-1:75000; Western Blot: 1:1000;

Type: Primary
Antigen: SMAD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse