Anti-IgG (H+L) Goat Polyclonal Antibody (Agarose)
Catalog # ROCK206-1150
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:Agarose
- Host:Goat
- ImmunoPrecipitation:Yes
- Reactivity:Guinea pig
- Cross adsorption:No
- Format:Antibody fraction
- Form:Suspension of agarose beads
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:+4 °C
- Shipping temperature:Wet Ice
- Size:20 mg
- Pk:20 mg
Specifications
About this item
Recommended Dilutions:
Do not freeze!
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Agarose
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Guinea Pig