Anti-Galectin-3 Rat Monoclonal Antibody [clone: M3/38]
Catalog # USBIG1044-53G-50
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Galectin 3
- Antigen symbol:GAL3
- Clonality:Monoclonal
- Clone:M3/38
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:–20 °C
- Concentration:~0,5 mg/ml
- Immunogen:Raised against mouse galectin-3
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: Galectin-3
Clonality: Monoclonal
Clone: M3/38
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human, Mouse,