Anti-ALIX Mouse Monoclonal Antibody [clone: 3A9]
Supplier: US Biological
Certificates
About this item
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: ALIX
Clonality: Monoclonal
Clone: 3A9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Frog, Human, Mouse,
Specifications
- Catalog No:
- USBIA1355-64D-25
- USBIA1355-64D-100
- Antigen symbol:
- ALIX
- ALIX
- Antigen name:
- Apoptosis-Linked Gene 2-Interacting Protein X
- Apoptosis-Linked Gene 2-Interacting Protein X
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 3A9
- 3A9
- Reactivity:
- Frog, Human, Mouse
- Frog, Human, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant corresponding to full-length human Alix.
- Recombinant corresponding to full-length human Alix.
- Form:
- liquid
- liquid
- Purification:
- purified by immunoaffinity chromatography
- purified by immunoaffinity chromatography
- Concentration:
- ~0,5 mg/ml
- ~0,5 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7,2, 0,09% sodium azide.
- PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:
- –20 °C
- –20 °C