Standard ground joints
Supplier: QUARTZTECHNISCHE WERKSTATTEN
Certificates
About this item
Connections, quartz glass, with standard ground joint according to DIN 12 249.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving high resistance to temperature changes.