Order Entry
Finland
ContactUsLinkComponent
Anti-eEF1B-beta1 and 2 Rabbit Polyclonal Antibody
Anti-eEF1B-beta1 and 2 Rabbit Polyclonal Antibody
Catalog # AGRIAS10677
Supplier:  Agrisera
undefined
Anti-eEF1B-beta1 and 2 Rabbit Polyclonal Antibody
Catalog # AGRIAS10677
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    eEF1B beta1/ 2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    A. thaliana
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    recombinant eEF1B-beta1 protein from Arabidopsis thaliana with no affinity tag, P48006, At1g30230
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: eEF1B beta1/ 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana