Anti-eEF1B-gamma1 and 2 Rabbit Polyclonal Antibody
Catalog # AGRIAS10676
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:eEF1B gamma1/2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombinant eEF1B-gamma1 protein from Arabidopsis thaliana with no affinity tag, O04487, At1g09640
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: eEF1B gamma1/2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana