Anti-IgE epsilon Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
AGRIAS101304
: Agrisera
- Antibody type:Secondary
- Antigen symbol:IgE epsilon
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Rabbit,Human,Bovine,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Storage temperature:+2°C to +8 °C
- Shipping temperature:Ambient
- Immunogen:Purified Human IgE (epsilon chain)
- Purification:purified
- Size:1 mg
- Pk:1 mg
Type: Secondary
Antigen: IgE epsilon
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rabbit