Order Entry
Finland
ContactUsLinkComponent
Anti-CRD1 Rabbit Polyclonal Antibody
Anti-CRD1 Rabbit Polyclonal Antibody
Catalog # AGRIAS06122
Supplier:  Agrisera
Anti-CRD1 Rabbit Polyclonal Antibody
Catalog # AGRIAS06122
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CRD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Chlamydomonas reinhardtii
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    residues 1-409 from Arabidopsis thaliana CHL27 fused to TrxA Q9M591, At3g56940
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: CRD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii