Anti-Cdk4 Rabbit Polyclonal Antibody
ORIGTA328198
: OriGene
- Antibody type:Primary
- Antigen symbol:CDK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rat
- Western blot:Yes
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Liquid
- Amino acid number:272 to 303
- Storage buffer:PBS with 0,09% (W/V) sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This Rat Cdk4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 272-303 amino acids from the C-terminal region of rat Cdk4.
- Purification:purified by Protein A chromatography, followed by affinity purification
- Size:400 μl
- Pk:400 µl
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: Cdk4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype:
Reactivity: Rat