Universal freezers, G models
Supplier: Liebherr
Certificates
About this item
These are general purpose freezers with static cooling - ideal for the storage of non critical products. Spacious, energy-saving, these upright freezers are very economical with optimised cooling systems and compressors. They use very little electricity and are very user friendly. All models are equipped with a recovery defrost water system. All models include SuperFrost technology which makes freezing simple and saves energy, it quickly reduces the temperature to −32 °C.
- Electronic temperature control
- All drawers can be removed to increase storage space
Temperature range: −18 to −25 °C