Dropping funnels, cylindrical, with pressure balance
Supplier: witeg Labortechnik
Certificates
About this item
Borosilicate glass, clear, with PTFE key.
- Cylindrical, graduated
- Drip-nozzle with pressure equalizer and screw-thread retaining nut
- With interchangeable valve made out of PTFE