Order Entry
Finland
ContactUsLinkComponent
Burner, Teclu
Burner, Teclu
Catalog # RLAA4010
Supplier:  R L ENTERPRISES
Burner, Teclu
Catalog # RLAA4010
Supplier:  R L ENTERPRISES
Specifications
  • Description:
    Teclu burner
  • Seq:
    01
  • Pk:
    1
  • For:
    Coal gas
Specifications
About this item
The burner is constructed with a gas control needle valve. Specially designed conical mixing chamber is incorporated, which allows the burner to give off a very hot flame.
  • Suitable for glass work and soldering
  • Gas and air mixing is regulated by adjusting the knurled wheel below the tube
Height: 145 mm