Order Entry
Finland
ContactUsLinkComponent
Dispenser bucket, Wiper Bowl® Safe & Clean
  115-2744
undefined
Dispenser bucket, Wiper Bowl® Safe & Clean
  115-2744
  • Description:
    Dispenser bucket, Wiper Bowl® Safe & Clean, empty
  • Pk:
    1
This dispenser bucket made of plastic can be filled with wipe rolls and disinfectant or cleaning solutions. It is ideal for thorough cleaning applications in sensitive areas, e.g. in workshops. If a work area is disinfected simply close the lid and the remaining cloths will remain moist until their next use - for 30 days.
  • Refillabl with off-the-shelf disinfectants or cleaning agents
  • Economic and hygienic
  • Easy dispensing thanks to tear-off perforation
Wipe rolls