Order Entry
Finland
Orders LinkContactUsLinkComponent
Bactident

Bactident

Bactident® Indole, 30 ml
Catalog # 1.11350.0001
Supplier:   Merck
Bactident
Catalog # 1.11350.0001
Supplier:  Merck
CAS Number:  
Bactident
Bactident

Bactident® Indole, 30 ml

CAS Number:  
Price
32,30 €
Log in for availability
1 * 30 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    30 ml
  • Description:
    Bactident® Indole

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.