Order Entry
Finland
ContactUsLinkComponent
Oxynex® LM, EMPROVE® ESSENTIAL antioxidant for fats and oils, suitable for use as excipient, SAFC®
Irritant
Oxynex® LM, EMPROVE® ESSENTIAL antioxidant for fats and oils, suitable for use as excipient, SAFC®
Catalog # 1.06946.1000
CAS Number:  
Irritant
Oxynex® LM, EMPROVE® ESSENTIAL antioxidant for fats and oils, suitable for use as excipient, SAFC®
Catalog # 1.06946.1000
CAS Number:  

Specifications

  • Pack type:
    Aluminium box
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • DL-α-Tocopherol (vitamin E) E 307 (HPLC):
    24.0 - 27.0 %
  • 6-Palmitoyl-L-ascorbic acid (ascorbyl palmitate) E 304 (HPLC):
    18.0 - 22.0 %
  • Identity:
    IR spectrum conforms
  • Al (Aluminium):
    ≤ 50 ppm
  • Ni (Nickel)*:
    ≤ 2 ppm
  • Sulfated ash:
    ≤ 2.5 %
  • Colony count (aerobic bacteria):
    ≤ 100 CFU/g
  • Colony count (Yeasts and moulds):
    ≤ 100 CFU/g
  • E.coli:
    absent in 1 g
  • Pseudomonas aeruginosa:
    absent in 1 g
  • Staphylococcus aureus:
    absent in 1 g
  • Candida albicans:
    absent in 1 g
  • Salmonella species:
    absent in 10 g
  • citric acid (calculated from weighing protocol):
    2.4 - 2.6 %
  • Lecithin E 322 (calculated from weighing protocol):
    24 - 25 %
  • Mono- and diglycerides of edible fatty acids E 471 (calculated from weighing protocol):
    26 - 27 %
  • Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).:
    24 - 25 %

Specification Test Results

About this item

 :
 :GHS07
Storage Temperature:Refrigerator