Order Entry
Finland
ContactUsLinkComponent
Parteck® ODT, EMPROVE® ESSENTIAL, SAFC®
Parteck® ODT, EMPROVE® ESSENTIAL, SAFC®
Catalog # 1.00490.9025
CAS Number:  
Parteck® ODT, EMPROVE® ESSENTIAL, SAFC®
Catalog # 1.00490.9025
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Identity:
    passes test
  • Appearance (description):
    fine powder
  • Appearance (color):
    white
  • Assay (mannitol):
    90.0 - 95.0 %
  • Assay (Croscarmellose-Sodium):
    3.0 - 7.0 %
  • Ni (Nickel)*:
    ≤ 0.0001 %
  • Heavy metals (as Pb):
    ≤ 0.0005 %
  • Ethanol (HS-GC):
    ≤ 5000 ppm
  • Other residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Water (according to Karl Fischer):
    ≤ 2 %
  • Microbiological purity (Total aerobic microbial count (TAMC)):
    ≤ 2000 CFU/g
  • Microbiological purity (Total yeast and mould count (TYMC)):
    ≤ 200 CFU/g
  • Bile tolerant Gram negative Bacteria (absent in 1 g):
    conforms
  • Escherichia coli (absent in 1 g):
    conforms
  • Pseudomonas aeruginosa (absent in 1 g):
    conforms
  • Staphylococcus aureus (absent in 1 g):
    conforms
  • Candida albicans (absent in 1 g):
    conforms
  • Salmonella ssp (absent in 10 g):
    conforms
  • Particle size (laser diffraction) (d10):
    30 - 90 µm
  • Particle size (laser diffraction) (d50):
    80 - 220 µm
  • Particle size (laser diffraction) (d90):
    170 - 520 µm

Specification Test Results

About this item

 :
Storage Temperature:Ambient