Order Entry
Finland
ContactUsLinkComponent
Culture media in sachets
Culture media in sachets
Catalog # SCAR01-111BA05
Supplier:  Scharlau
CAS Number:  
Culture media in sachets
Catalog # SCAR01-111BA05
Supplier:  Scharlau
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    Malt extract agar No. 1
  • Sterile water to be added:
    500 ml
Specifications
About this item
Dehydrated media in sachets are an ideal solution for media used in smaller quantities.
  • Fast
  • Simple
  • Safe
  • Less waste