Water treatment units: Acqua purificator G 7895/1
Supplier: MIELE
Certificates
About this item
Water treatment units for use with G 78XX series machines. Cabinet for two E310/318 water demineralisation cartridges with conductivity meter, generally recommended quality for final rinse <15 µs/cm.
- Two pressure hoses, approx. 1,2 m long, with ³/₄" threated union
- 1× cold water connection to cartridge, 2,5 to 10 bar pressure
- 1× connection from cartridge to machine (loss of pressure approx. 1 bar for each cartridge)
Casing made from white steel or stainless steel.