ESD safety shoes, slip-on, HACCP, Uni6, 31740
Supplier: Abeba
Certificates
About this item
ESD safety shoe made from microfibre upper, with non slip TPU walking sole, which is resistant to animal fats, oils and fuels.
- Perspiration-absorbent special lining with silver fibre
- Replaceable acc Wave insole, orthopaedic insoles possible
- Sanitized® treatment reduces odour and bacterial growth
- Shock-absorbent foam PU midsole
- Protective steel toecap (200 J)
- Washable at 30 °C
Suitable for hygienically demanding areas.
Colour: White
Certifications: EN 61340 (ESD); DGUV 112-191; HACCP; EN ISO 20345:2011; S2, SRA