Anti-EP4 receptor C-terminal Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # COBSD5-1866-50
Supplier: Columbia Biosciences
Specifications
- Antibody type:Primary
- Antigen name:EP4 receptor
- Clonality:Polyclonal
- Conjugation:RPE (R-Phycoerythrin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: EP4 receptor
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: