Order Entry
Finland
ContactUsLinkComponent
Anti-EP2 receptor Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Anti-EP2 receptor Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
  COBSD5-1863-50
Anti-EP2 receptor Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
  COBSD5-1863-50

 

  • Antibody type:
    Primary
  • Antigen name:
    EP2 receptor
  • Clonality:
    Polyclonal
  • Conjugation:
    RPE (R-Phycoerythrin)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Amino acid number:
    335 to 358
  • Sequence:
    SLRTQDATQTSCSTQSDASKQADL
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: EP2 receptor
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: