Anti-6X His tag Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: AD1.1.10]
COBSD11-1711
- Antibody type:Tag
- Antigen name:egl-9 family hypoxia-inducible factor 2
- Antigen symbol:EGLN2
- Clonality:Monoclonal
- Clone:AD1.1.10
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:none
- Gene ID:112398
- Antigen synonyms:HIFPH1|HIF-PH1|EIT6|HPH-1|HPH-3|PHD1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: monoclonal
Clone: AD1.1.10
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: