Order Entry
Finland
ContactUsLinkComponent
Anti-IgA alpha Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-IgA alpha Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # IMMRIR1534
Supplier:  ImmunoReagents
Anti-IgA alpha Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # IMMRIR1534
Supplier:  ImmunoReagents

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin A alpha
  • Antigen symbol:
    IgA
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgA
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Clear, fluorescent yellow liquid
  • Storage buffer:
    10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
  • Storage temperature:
    2…8 °C
  • Concentration:
    1,50 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2…8 °C
  • Immunogen:
    Human IgA, (α chain)
  • Tested applications:
    Fluoresence ELISA, Fluorescence-Activated Cell Sorting
  • Purification:
    Affinity purified using solid phase Human IgA
  • Size:
    1,0 mg
  • Pk:
    1 mg

Specifications

About this item

ImmunoReagents FITC Conjugates are used in immunofluorescence microscopy, flow cytometry, FLISA (fluorescent ELISA) and high throughput screening assays. Fluorescein is typically excited by the 488 nm line of an argon laser, and emission is collected at 518 nm. FITC antibody conjugates are yellow-orange in color but emit a yellow-green color upon Em 518 nm. FITC antibody conjugates are sensitive to light. The typical FITC to antibody ratio (F/P) is >4.0 Excitation/Emission = 494 nm / 518 nm Emission Color = Green (Similar Dyes: Alexa Fluor 488, Cy2, DyLight ® 488).

  • Origin: US

ImmunoReagents antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.
Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgA alpha
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgA
Reactivity: Human