Anti-VEGFA Mouse Monoclonal Antibody (Biotin) [clone: 0.N.600]
USBIV2110-13A
- Antibody type:Primary
- Antigen name:Vascular endothelial growth factor A
- Antigen symbol:VEGFA
- Clonality:Monoclonal
- Clone:0.N.600
- Conjugation:Biotin
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Rabbit,Human
- Neutralisation:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Concentration:~0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µl
- Pk:500 µl
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: monoclonal
Clone: 0.N.600
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rabbit