Anti-SORCS3 Mouse Monoclonal Antibody [clone: 9G445]
USBIS5354
- Antibody type:Primary
- Antigen name:sortilin-related VPS10 domain containing receptor 3
- Antigen symbol:SORCS3
- Clonality:Monoclonal
- Clone:9G445
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:1147 to 1222
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: SORCS3 (sortilin-related VPS10 domain containing receptor 3)
Clonality: Monoclonal
Clone: 9G445
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human