Anti-SIRP, alpha1 Mouse Monoclonal Antibody (Biotin) [clone: 3H1939]
USBIS1013-87P
- Antibody type:Primary
- Antigen symbol:SIRP, alpha1
- Clonality:Monoclonal
- Clone:3H1939
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Rat
- Form:liquid
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: SIRP, alpha1
Clonality: Monoclonal
Clone: 3H1939
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat