Anti-Semaphorin 5A Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIS0668-57B-BIOT
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Semaphorin 5A
- Antigen symbol:SEMA5A
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: Semaphorin 5A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse