Anti-Reg IIIa Goat Polyclonal Antibody
Catalog # USBIR1374-80
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Reg IIIa
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:27 to 175
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,2 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.1 ?g/ml
Type: Primary
Antigen: Reg IIIa
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat