Anti-POU Domain, Class 3, Transcription Factor 2 Goat Polyclonal Antibody
Catalog # USBIP5250-23C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:POU Domain, Class 3, Transcription Factor 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Pig,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
- Sequence:C-AQSLVQGDYGALQSN
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:8000; Western Blot: 0.1-0. 3?g/ml
Type: Primary
Antigen: POU Domain, Class 3, Transcription Factor 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Pig, Rat