Anti-Plasmodium falciparum, Schizont Infected Red Blood Cells Mouse Monoclonal Antibody [clone: 10F298]
Catalog # USBIP4256-70B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Plasmodium falciparum, Schizont Infected Red Blood Cells
- Clonality:Monoclonal
- Clone:10F298
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgM
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Plasmodium falciparum, Schizont Infected Red Blood Cells
Clonality: monoclonal
Clone: 10F298
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: