Anti-Phospholipase C, epsilon Rabbit Polyclonal Antibody
Catalog # USBIP4074-41R
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Phospholipase C, epsilon
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:2285 to 2303
- Storage buffer:0,1M Tris-glycine, pH 7,4, 0,15M sodium chloride, 0,05% sodium azide, before the addition of glycerol to 30%.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:250-1:1000
Type: Primary
Antigen: Phospholipase C, epsilon
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human