Anti-SLC12A2 Rabbit Polyclonal Antibody
Catalog # USBIN2800-01B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 12 (sodium/potassium/chloride transporter), member 2
- Antigen symbol:Solute carrier family 12, member 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage buffer:10mMsodium HEPES pH 7,5, 150mM NaCl, 0,1mg/ml BSA, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: SLC12A2 (solute carrier family 12 (sodium/potassium/chloride transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human