Anti-Gravin Goat Polyclonal Antibody
Catalog # USBIG8959-81C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Gravin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris-saline, 0,02% sodium azide, 0,5% BSA.
- Sequence:C-KQERESAKSELTES
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Gravin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human