Order Entry
Finland
ContactUsLinkComponent
 
Anti-FGFR2 Rabbit Polyclonal Antibody
  USBIF4305-16G
undefined
Anti-FGFR2 Rabbit Polyclonal Antibody
  USBIF4305-16G

 

  • Antibody type:
    Primary
  • Antigen name:
    Fibroblast growth factor receptor 2
  • Antigen symbol:
    FGFR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Form:
    liquid
  • Amino acid number:
    15 to 44
  • Storage buffer:
    PBS, pH 7,4, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: FGFR2 (fibroblast growth factor receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human