Anti-Eosinophil Major Basic Protein Mouse monoclonal antibody [clone: BMK-13]
Catalog # USBIE3330-01D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Eosinophil Major Basic Protein
- Clonality:Monoclonal
- Clone:BMK-13
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,1% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: Immunochemistry: 1:30
Type: Primary
Antigen: Eosinophil Major Basic Protein
Clonality: monoclonal
Clone: BMK-13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human