Order Entry
Finland
ContactUsLinkComponent
Anti-Collapsin Response Mediator Protein 4 Rabbit Polyclonal Antibody
Anti-Collapsin Response Mediator Protein 4 Rabbit Polyclonal Antibody
Catalog # USBIC7517-08K
Supplier:  US Biological
Anti-Collapsin Response Mediator Protein 4 Rabbit Polyclonal Antibody
Catalog # USBIC7517-08K
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Collapsin Response Mediator Protein 4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Monkey,
    Human,
    Rat,
    Mouse,
    Cat
  • Western blot:
    Yes
  • Format:
    Serum
  • Form:
    liquid
  • Sequence:
    YDGPVFDLTTTPK
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: Collapsin Response Mediator Protein 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Cat, Human, Monkey, Mouse, Rat