Anti-Collapsin Response Mediator Protein 4 Mouse Monoclonal Antibody [clone: 9G340]
Catalog # USBIC7517-08H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Collapsin Response Mediator Protein 4
- Clonality:Monoclonal
- Clone:9G340
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:457 to 556
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Collapsin Response Mediator Protein 4
Clonality: Monoclonal
Clone: 9G340
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat