Anti-Collapsin Response Mediator Protein 3 Mouse Polyclonal Antibody
USBIC7517-07C
- Antibody type:Primary
- Antigen name:Collapsin Response Mediator Protein 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Format:Serum
- Form:liquid
- Storage buffer:50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: Collapsin Response Mediator Protein 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human