Order Entry
Finland
ContactUsLinkComponent
Anti-CD3, epsilon Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K105]
Anti-CD3, epsilon Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K105]
Catalog # USBIC2254-19H
Supplier:  US Biological
Anti-CD3, epsilon Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K105]
Catalog # USBIC2254-19H
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD3 epsilon
  • Clonality:
    Monoclonal
  • Clone:
    5K105
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Pig
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,1% sodium azide
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

Type: Primary
Antigen: CD3, epsilon
Clonality: Monoclonal
Clone: 5K105
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Pig