Anti-CD3, epsilon Mouse Monoclonal Antibody [clone: 5K105]
Catalog # USBIC2254-19G
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CD3 epsilon
- Clonality:Monoclonal
- Clone:5K105
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Pig
- Environmentally Preferable:
- Form:liquid
- Storage buffer:100mM borate buffered saline, pH 8.0. No oramine- containing buffer salts .
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: CD3, epsilon
Clonality: Monoclonal
Clone: 5K105
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Pig