Anti-CD3, epsilon Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5K4]
Catalog # USBIC2254-11Q
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CD3 epsilon
- Clonality:Monoclonal
- Clone:5K4
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2c kappa
- Reactivity:Mouse
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide and a stabilizing agent.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: CD3, epsilon
Clonality: Monoclonal
Clone: 5K4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2c Kappa
Reactivity: Mouse