Anti-CD3, epsilon Hamster Monoclonal Antibody (Cy5®)) [clone: 5K3]
Catalog # USBIC2254-11K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CD3 epsilon
- Clonality:Monoclonal
- Clone:5K3
- Conjugation:Cy5®
- Flow cytometry:Yes
- Host:Hamster
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Mouse
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CD3, epsilon
Clonality: Monoclonal
Clone: 5K3
Conjugation: Cy5®
Epitope:
Host: Hamster
Isotype: IgG Kappa
Reactivity: Mouse