- Antibody type:Primary
- Antigen symbol:CD3 epsilon
- Clonality:Monoclonal
- Clone:5K3
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Hamster
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Mouse
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide and a stabilizing agent.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Do not freeze!
Type: Primary
Antigen: CD3, epsilon
Clonality: Monoclonal
Clone: 5K3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Hamster
Isotype: IgG Kappa
Reactivity: Mouse